Rabbit NIH-32 Open Formula, Autoclavable, Zeigler™
Supplier: Zeigler
Certificates
About this item
Autoclavable diet designed for maintenance, growth, and reproduction of rabbits
- Contains higher concentrations of vitamins to compensate for losses that occur during steam sterilization
- Contains 14% protein, 2.5% fat
- Pellet size: 4 mm (5/32")
Ingredients include: ground barley, soybean meal, oat hulls, wheat middlings, wheat bran, dried whey, alfalfa meal, brewer’s dried yeast, soy oil, dicalcium phosphate, limestone, salt, and premixes.
Max. ash: 10%
Max. crude fibre: 22%
Certifications: Used as a standard reference diet in rabbit colonies involved in biological and biomedical research.
Customers Who Bought This Also Bought