Portable Digital Field Scale
Catalog #: 470356-296
Supplier: AEROLITE METEORITES INC SE
New Product
Specifications
- Description:Portable digital field scale
- Weighing Capacity:600 g
Specifications
About this item
This remarkable little scale is sturdy and easy to use.
- Six units: g, oz., gn, ozt, dwt, ct
- Two easy to use buttons
- LCD with blacklight
- Easy calibration
A good digital scale is indipensable, both in the field and collection room.
Accuracy: ±0.1
Customers Who Bought This Also Bought