About this item
Spor-Klenz® fast-acting, liquid cold sterilant/disinfectant requires no mixing or activation
Spor-Klenz® fast-acting, liquid cold sterilant/disinfectant requires no mixing or activation
Use wherever critically high-level microbial control is required. Ready-to-use format eliminates errors in dilution preparation. Low-toxicity profile adds to worker safety. Undiluted, the product is sporicidal in 51/2 hours and disinfects in 10 minutes. Accompanied by certificates of analysis, filtration, and sterility.
A fast acting, liquid cold sterilant/disinfectant, filtered to 0.22 micron and specifically formulated for use in the sterilization and disinfection of hard environmental surfaces in Pharmaceutical, Medical Device, Biotech and Cosmetic manufacturing facilities. This product is a stabilized blend of peracetic acid, hydrogen peroxide, and acetic that provides fast, effective microbial control, including spores.