TRIO.BAS™ SELFTEST Base Station Induction Battery Charger Calibration Check System
Supplier: Hardy Diagnostics
Certificates
About this item
The Base Station Induction Battery Charger with user Self-test is equipped with a system that allows users to check the state of precision of the airflow, regardless of auto-calibration already present in the instrument. This verification is necessary to avoid the possible invalidation of the tests between annual controls for official certification.
- Calibration bell
- Connector cable and charger with outlet plug
This base is used in replacement of the base station induction charger when the Self-test system is applied.
Note: this unit does not take the place of official calibration services as outlined by the USP.
Specifications
- Catalog No:
- 75804-762
- 75804-760
- Flow rate:
- 200 L/min
- 100 L/min