Vee Gee Filter Holder Components
Supplier: Heathrow Scientific
Certificates
About this item
Available with fritted glass support base for general and biological applications, or with removable stainless steel screen support base and PTFE gasket for ultraclean filtrate or proteinaceous solutions.
- For use with Vee Gee Filter system kits
All items except the PTFE gasket are autoclavable.
Customers Who Bought This Also Bought