Millennium Citrus Cleaners, CANI
Supplier: Cani
Certificates
About this item
Highly concentrated cleaners designed to cut through the toughest soil conditions, and leave surfaces sparkling clean with a fresh, pleasant citrus odor
Cleaner is available in general-purpose and low-foaming formulations. The general-purpose cleaners contain an effective buffering agent combined with natural citrus oils to maintain cleaning strength under heavy soil loads. Cleaners also contain a fast-acting, powerful solubilizing agent that dissolves and suspends any soil, allowing it to be wiped or rinsed away completely. The low-foaming cleaners can be used as a detergent additive in immersion, pressure washing, and circulation cleaning for the cosmetic, pharmaceutical, medical device, and food and beverage industries.
Customers Who Bought This Also Bought