Synthware Short Path Distillation Apparatus with [ST]24/40 Joint, Kemtech America
Catalog #: 60002-078
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Cat. no.:60002-078
- ST Joint Size:24/40
- Thermometer ST Joint Size:10/40
- Supplier No.:D261024
Specifications
About this item
One-piece design for easy set up.
- Heavy wall borosilicate construction
- Short path
- Ground glass joint
- Jacketed
- Designated 10/18 thermometer joint
Used with 25 mm immersion thermometer.
Ordering information: Distillation receiver and thermometer are not supplied. For best results, use distillation receiver 60002-064. For thermometer, contact your Avantor representative.
Customers Who Bought This Also Bought