VWR® Safetypette™ Pipette Filler
Supplier: VWR International
CE Compliant
Certificates
About this item
Designed to reduce the strain associated with serial pipetting while providing simple, precise operation for right- and left-handed users.
- Reduces strain
- Ambidextrous
- Lightweight
Compatible with 1 to 100 ml blow-out and 'to deliver' pipettes.
As much as 65 ml can be aspirated from a single squeeze of the bulb. Aspiration and dispensation speed is precisely controlled with the simple thumb lever. A mini squeeze bulb evacuates blow out pipettes. Silicone pipette holder is washable and autoclavable.
Weighs only 102 g, sits comfortably in either hand.
Ordering information: Supplied with replaceable integral 0.45 µm filter and mint, white, pink, and mauve cones for color coding. Three-year manufacturer's warranty also included.
Customers Who Bought This Also Bought