KONTES® Distillation Apparatus, Short Path, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Design utilizes a thermometer bulb as column packing
Compact head and condenser provide better separation by reducing wetted surfaces. “Cow” type receiver has tubes that are aligned for best product recovery. Pear shaped flask has [ST] 10/18 side arm for bleed tube. 14/20 [ST] joints. #1 Hose connections. 190Hx215Wmm.
Customers Who Bought This Also Bought