Synthware Distillation Apparatus, Large Scale, Kemtech America
Catalog #: 60002-152
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Cat. no.:60002-152
- ST Joint Size:24/40
- Supplier No.:D502440
Specifications
About this item
One-piece design with [ST]10/18 thermometer joint and high-efficiency cooling jacket
- Heavy wall borosilicate construction
- Ground glass joint
- Jacketed
- Large scale applications
Used with 50mm immersion thermometer.
Ordering information: Thermometer is not supplied; contact your Avantor representative for more information.
Customers Who Bought This Also Bought