WHEATON® E-Z Crimper™ Hand-Operated Crimpers, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Crimpers feature polished crimping jaws for consistent sealing, a redesigned spring assembly, and a short, cushioned ergonomic handle that helps reduce hand fatigue and adds comfort
Note: Autoclaving may change the appearance or reduce product life.
An adjustable stop allows tightening of the seal while maintaining consistent crimping. The 8, 11, and 13mm models feature tapered jaws for easier vial access in rack. Each crimper size is labeled for quick identification. Autoclavable at 121°C (250°F).
Ordering information: Crimpers work with the Crimpmaster® crimping station. Crimper 80078-992 is supplied with an O-ring to provide the option of crimping the top of a vial to create an extra tight seal. This option is recommended only for standard-opening vials.
Related Information
Certificates
Search by Lot NumberDWK Life Sciences Return to Research Promo 2021
Promotion Material
Wheaton Vial GUide
Product Literature
Customers Who Bought This Also Bought