Specifications
- Dimensions:13.5×5.75×1.5"
- Description:Upgraded retort stand set
- Weight:4.2 lbs.
Specifications
About this item
The most compact retort stand, rod, clamp and ring set on the market!
- Durable construction
- Four piece set
This comprehensive set now features a two-piece stainless steel rod that effortlessly screws together, ensuring easy setup and enabling more compact storage when not in use. The retort stand is made of corrosion and scaling resistant, powder coated pressed steel, and features a tapped hole, suitable for connecting the included rod, or any other rods with a 10×1.5 mm metric thread. The stand measures 8" long, 5" wide and is approximately 1" in height. Included rod is made of stainless steel and measures 23.6" tall, 10 mm in diameter and features a 10×1.5 mm metric thread. The ring features a zinc plated steel shank secured to a screw clamp. Distance from each of the rings centers to the center of the support measures 4.75". Clamp is four-pronged with a cork lining.