Acurex™ 501 and 502 Compact Bottle-Top Dispensers, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Compact dispensers for safe reagent handling, adapted to refrigerator storage as well as water bath heating
Dosing mechanism entirely protected inside reservoir. Manufactured and tested to fully comply with the latest regulations on instrument safety and precision. Volumes range from 0.2 to 30 mL. This product is also fully autoclavable at 121°C (250°F).
Ordering information: One-year manufacturer's warranty.
Customers Who Bought This Also Bought