Cannon-Fenske Routine Viscometers, Calibrated, Technical Glass Products
Supplier: Avantor
Certificates
About this item
For use in the petroleum and chemical industries
Viscosities can be measured with a precision of ±0.2% quickly and easily. Require minimum sample of approximately 7mL. Specifications conform to ASTM Standard D446. Calibrated by manufacturer and supplied with certificate. Chemically-resistant plastic holder (sold separately) slips over the viscometer and keeps it firmly in place with a spring clip.
Customers Who Bought This Also Bought