About this item
Eppendorf's twin.tec® 96-well, 150 µl PCR Plates feature a polycarbonate frame and thin-walled polypropylene wells ensuring optimal heat transfer and high rigidity. The SafeCode variants enhance sample identification, while the BioBased options, manufactured using renewable resources, provide an eco-friendly alternative without compromising performance.
- One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance
- Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample
- Exceptionally solid and torque-resistant polycarbonate frame
- Improved well-to-well tolerance
- Cut-away corner and alphanumeric grid-referencing
- Raised well rims for effective sealing, also reduces risk of cross-contamination
- Batch-tested and certified free of DNA, DNase, RNase and PCR inhibitors (PCR clean)
The Eppendorf twin.tec® PCR plate combines extremely thin-walled polypropylene wells for optimal heat transfer to the sample, with robust polycarbonate frames for high rigidity. With raised well rims, twin.tec plates enable highly effective sealing to reduce the risk of cross-contamination.
Twin.tec plates are available in many different formats to suit different workflows and applications, for example: skirted, semi-skirted, and unskirted; divisible plates to avoid waste in small assays; individually wrapped forensic DNA grade plates; microbiology plates; and barcoded plates.
All Eppendorf twin.tec PCR plates are ‘PCR clean’, having been batch-tested and certified by an independent laboratory to be free of a range of common PCR contaminants, including DNA, DNAse, RNAse, and PCR inhibitors. With twin.tec PCR plates you’ll have maximum confidence in your results.
BioBased (made of bio-based material) varient aids in the reduction of consumable-related carbon footprint in the lab. The bio-based plates have identical material properties and technical specifications as our non-BioBased Plates.
Barcoded option provides 3-level coding for safe sample and vessel identification. ECC 200 error correction function enables safe code reading with up to 30 % damaged codes.
Autoclavable: 121 °C, 20 min
Centrifugation stability: 2250 ×g
Specifications
- Catalog No:
- CA35920-176
- CA95041-434
- CA95041-432
- CA95041-430
- CA89136-702
- CA89136-680
- CA35920-172
- CA35920-170
- CA95041-438
- CA95041-436
- CA89136-704
- CA89136-686
- CA89136-692
- CA35920-178
- CA89136-690
- CA35920-174
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001