VWR® Talon® Large Lab-Frames
Supplier: VWR International
Certificates
About this item
These 1220x1220mm (48x48") frames are ideal for complex glassware setups.
- For large-capacity setups
- Stable
- Ideal for complex glass setups
Base is 460mm (18") wide for stability.
Ordering information: Each Lab Frame includes ten 51 mm (2") rods, three 457 mm (18") rods, two 914 mm (36") rods, ten 1220 mm (48") rods, four rod end connectors, thirty-eight S-connectors, and six Lab Frame feet.
Customers Who Bought This Also Bought