VWR® Talon® Medium Lab-Frames
Supplier: VWR International
Certificates
About this item
Lab Frames are convenient for distillation and general setups.
- For horizontal or vertical mounting
- Convenient size for distillation and general setups
- Stable base
Can be assembled as 1220Lx610Hmm (48x24") horizontal or 610Lx1220Hmm (24x48") vertical frame. Base is 46 cm (18") wide for stability.
Ordering information: Each Lab Frame includes eight 51 mm (2") rods, two 457 mm (18") rods, seven 610 mm (24") rods, four 1220 mm (48") rods, four rod end connectors, thirty-five S-connectors, and four Lab Frame feet.
Customers Who Bought This Also Bought