Orders LinkContactUsLinkComponent
Chemdye® Three Line Automatic Labeler
Automatic labeler
  77506-952
 :   TERRAGENE
 :  CG3
Chemdye® Three Line Automatic Labeler
  77506-952
 :  TERRAGENE
Chemdye® Three Line Automatic Labeler
Chemdye® Three Line Automatic Labeler
Automatic labeler
$1,046.91
Each
Three line labeler for quick and easy labeling of sterilization package with Chemdye® CD13/ CD23/ CD33/ CD43/ CD53/ CD73/ CD203/ CD403 labels.
  • 3-Line labeler has 3 print lines with 12 alphanumeric characters each
  • Allows printing alphanumeric data on Chemdye® labels
  • Enabling easy and clear traceability of packages to be sterilized
On each label, the following information can be printed: sterilizer number, batch, sterilization date, initials of the person responsible, package content, expiration date or other information. These labels have a Type 1 chemical indicator that changes color in the presence of the sterilizing agent and allows distinguishing processed packages from unprocessed ones.
With this labeler you can print all information needed for traceability and release of a sterilization batch, providing an effective way to document the sterile material to be used.
 : 1 unit per box.
  • Description:
    Automatic labeler