PicoSolutions Alternative Energy Education Lab Plus Kit
Supplier: PICOSOLUTIONS GROUP INC SE
New Product
Certificates
About this item
Explore alternative energy with a DIY hands-on turbine model.
- 3rd grade to college level tailored activities for different learning stages
- Includes DC turbines (horizontal and vertical) and a 7.2 V solar panel for a full spectrum of alternative energy exploration
- Versatile configuration support enables both AC and DC configurations
- Subjects spanning sustainable energy, math, aerodynamics, geometry
Build and test a wind turbine kit contents includes vertical and horizontal turbine level 1 kits, six hydro paddles, a multimeter, and a 7.2 V 200 mA solar panel.
Compared to the basic kit (470355-302) the plus kit adds more features, including all our turbines (horizontal, vertical, and hydro), a 7.2 V solar panel, and comprehensive electricity labs.
Ordering information: DESS course upgrade: Enhance learning with an upgrade to our design entrepreneurship and STEM skills (DESS) course, designed to cultivate creativity, innovation, and problem-solving skills, preparing students for a future in technology and engineering.
Specifications
- Catalog No:
- 470355-354
- 470355-374
- 470355-358
- 470355-388
- 470355-326
- 470355-370
- Dimensions:
- 17×16×8"
- 17×16×8"
- 17×16×8"
- 17×16×8"
- 17×16×8"
- 17×16×8"