PicoSolutions Vertical Wind Turbine Plus Kit
Supplier: PICOSOLUTIONS GROUP INC SE
New Product
Certificates
About this item
Explore vertical wind energy with a DIY hands-on tubine model.
- 6th grade to college level tailored activities for different learning stages
- Subjects spanning sustainable energy, math, aerodynamics, geometry
- Easy 10 minute assembly
Build and test a wind turbine to teach the power of the wind the kit students to experiment with wind power using DC and AC configurations and showcasing the power with an LED, a piezo buzzer, or a multimeter. The kit uses an LED, a buzzer, or a multimeter to show how wind generates electricity. The kit includes all parts for building the turbine.
Compared to the standard model (470355-312), the plus version includes the AC generator with magnets, coils, and over 12 electricity labs.
Ordering information: DESS course enhancement: Offers an upgrade to a specialized course that melds design, entrepreneurship, and STEM, aimed at cultivating creativity, innovation, and problem-solving prowess.
Specifications
- Catalog No:
- 470355-314
- 470355-356
- 470355-362
- 470355-352
- 470355-324
- 470355-378
- Dimensions:
- 11×8×6"
- 11×8×6"
- 11×8×6"
- 11×8×6"
- 11×8×6"
- 11×8×6"