PicoSolutions Horizontal Wind Energy Plus Kit
Supplier: PICOSOLUTIONS GROUP INC SE
New Product
Certificates
About this item
Explore horizontal wind energy with a DIY hands-on turbine model, now including AC and DC power generation.
- 3rd grade to college level tailored activities for different learning stages
- Subjects spanning sustainable energy, math, aerodynamics, geometry
- Easy 10 minute assembly
Build and test a wind turbine to teach the power of the wind Features a DC motor for beginners and an AC generator with 12+ experiments for advanced learners. The kit uses an LED, a buzzer, or a multimeter to show how wind generates electricity. The kit includes all parts for building the turbine, with blade adapters for custom blades made from dowels or 3D printing.
In comparsion to the standard kit (470355-376), the plus kit includes an AC generator with 12+ experiments for advanced learners.
Ordering information: Educational support bundle: Upgrade available for enhanced learning with support from a certified virtual teacher mentor and DESS course.
Future-ready skills: The DESS course enriches the kit by integrating design, entrepreneurship, and STEM, fostering creativity, innovation, and problem-solving abilities.
Future-ready skills: The DESS course enriches the kit by integrating design, entrepreneurship, and STEM, fostering creativity, innovation, and problem-solving abilities.
Specifications
- Catalog No:
- 470355-344
- 470355-386
- 470355-298
- 470355-306
- 470355-338
- 470355-392
- Dimensions:
- 17×16×8"
- 17×16×8"
- 17×16×8"
- 17×16×8"
- 17×16×8"
- 17×16×8"