Hastings Triplet Magnifiers, Bausch & Lomb®
Supplier: Avantor
Certificates
About this item
Highly corrected aplanatic magnifiers have three separate lenses, bonded together to form a compound lens
Provide sharp, very distinct images, with no color distortion. Swing-away nickel plated case.
Customers Who Bought This Also Bought