Coddington Magnifiers, Bausch & Lomb®
Supplier: Avantor
Certificates
About this item
A single lens constructed with an internal diaphragm
Good definition and a wide field. Mounted in a swing-away nickel-plated case.
Customers Who Bought This Also Bought