Black Polypropylene Screw Cap for Vials, PTFE/Silicone Liner, WHEATON®
Catalog #: 89050-868
Supplier: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Specifications
- Cap Size:8-425
- Cat. no.:89050-868
- Supplier No.:240602
Specifications
About this item
Cap is compatible with 12x32mm standard opening screw top vials for autosampling.
Customers Who Bought This Also Bought