EP Scientific Polypropylene Open-Top Screw Cap with PTFE/Silicone Septum, Thermo Scientific
Catalog #: 89039-866
Supplier: Thermo Scientific
Specifications
- Cap Size:20-400
- Septum:Bonded Resealable PTFE-lined Silicone Septa
- Color:White
- Description:Open-Top Septa Cap
- Cat. no.:89039-866
- Supplier No.:20-400/WS-3
Specifications
About this item
For use with EP Scientific EPA Vials
Cap has welded PTFE/silicone resealable septum. Cap is not pre-cleaned and is ready for your own cleaning procedure.
Customers Who Bought This Also Bought