DuPont™ ProClean® Coveralls
Supplier: DuPont
Certificates
About this item
ProClean® offers improved non-hazardous light liquid splash protection.
- Serged seams
- Bound neck
- Set sleeve design
- Covered elastic wrists and ankles
- Zipper closure
- Enclosed elastic at back
ProClean® offers improved non-hazardous light liquid splash protection.
ProClean® is made from a microporous composite fabric. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Coveralls feature covered elastic openings at wrists and ankles for a tighter fit. Bound neck ensures lower particle shedding. Front zipper closure allows for easy donning and doffing. Enclosed elastic at the back of the garment allows for a better fit.
Packaging: Bulk packaged garments are bulk packaged in double transparent poly liners.
Specifications
- Catalog No:
- CA13451-196
- CA13451-206
- CA13451-208
- CA13451-202
- CA13451-204
- Garment application:
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory