DuPont™ ProShield® NexGen® Coveralls
Catalog #: CA13451-026
Supplier: DuPont
Specifications
- Clothing Size:3XL
- Garment application:Laboratory
- Color:White
- Description:Elastic Wrists and Ankles, with Attached Hood
- Cat. no.:CA13451-026
- Supplier No.:NG127SWH3X00
Specifications
About this item
Economical, disposable, polypropylene coveralls offer protection and comfort in nonhazardous workplace environments.
- Serged seams
- Standard hood with elastic face opening
- Skid-resistant material
Economical, disposable, polypropylene coveralls offer protection and comfort in nonhazardous workplace environments.
A proprietary microporous technology provides an effective barrier against a variety of common industrial aerosols and liquids. Ideal for situations where dry particulate protection and liquid repellency are needed. Antistatic treated. Available in four styles, all with serged seams and front zipper closure.
Packaging: Bulk packed.
Customers Who Bought This Also Bought