KONTES® Needle Spargers, Purge and Trap, Kimble Chase
Catalog #: KT591150-0005
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Capacity:5 ml
- Cat. no.:KT591150-0005
- Supplier No.:591150-0005
Specifications
About this item
Designed for use with Tekmar® 2000 and 3000 purge and trap concentrators and 2016 and 2032 autosamplers for analysis of slurries, soils, viscous liquids, and foaming liquid samples.
These spargers are also used for headspace sweep analysis of volatile compounds in oils.
Customers Who Bought This Also Bought