KONTES® U-Shaped Spargers, Fritted, for Vial Autosamplers, Kimble Chase
Catalog #: KT591105-2605
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Capacity:5 ml
- Description:Fritted Sparger with Left Stem
- Cat. no.:KT591105-2605
- Supplier No.:591105-2605
Specifications
About this item
Designed for use with the Tekmar® purge and trap concentrators when using either the AQUATek 50 or ALS 2050 vial autosamplers
These fritted spargers are available with the stem on either the right or left side to provide the greatest flexibility in equipment layout. The internal standard vessel allows a visual check of the remaining standard available for use.
Customers Who Bought This Also Bought