VWR® Signature™ Shoe Covers made with DuPont™ Tyvek® IsoClean® Material
Supplier: VWR International
Certificates
About this item
Shoe covers are manufactured using a unique flash spinning process that creates an excellent barrier against nonhazardous liquids and microorganisms as well as dry particles down to 0.5µm.
- Excellent barrier against nonhazardous liquids and microorganisms, dry particles down to 0.5µm
- For cleanroom use, chemically and biologically inert, tear resistant, and low linting
- Single use, serged seams and elastic tops for a secure fit
- Gripper™ soles provide enhanced skid-resistance and durability
Shoe covers are manufactured using a unique flash spinning process that creates an excellent barrier against nonhazardous liquids and microorganisms as well as dry particles down to 0.5µm.
Designed for cleanroom use, garments are chemically and biologically inert, tear-resistant, and low-linting. Single use shoe covers feature serged seams and elastic tops for a secure fit. Also equipped with flat, non-skid PVC soles.
Ordering information: Bulk packed.
Specifications
- Catalog No:
- 89128-790
- 89128-792
- 89128-794
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom
Specifications
Related Information
Certificates
Search by Lot NumberFocus Safety Volume 1 Issue 1 Safety Essentials Volume 2
Product Literature
Customers Who Bought This Also Bought