ContactUsLinkComponent
Silver Shield®/4H® Booties, Honeywell Safety
Silver Shield®/4H® Booties, Honeywell Safety
Catalog #: CA11000-629
Booties
Silver Shield®/4H® Booties, Honeywell Safety
Catalog #: CA11000-629
Specifications
  • Garment application:
    Chemical protection
  • Shoe size:
    Universal
  • Description:
    Silver shield®/4H® Booties
  • Supplier No.:
    SSB
Specifications
About this item
Booties feature a five-layer fabric compacted into 2.7 mil
  • Five-layer fabric compacted into 2.7 mil
  • High-energy polymer (EVOH) with a low-energy polymer (PE), impervious to at least 90% of all organic chemicals for 4 hours at 35 °C (95 °F)
  • Each layer acts as a barrier against various epoxies and industrial chemicals
  • Wear as a liner or as a cover
  • One size fits all
Laminating a high energy polymer (EVOH) with a low-energy polymer (PE) creates a fabric impervious to at least 90% of all organic chemicals for four hours at 35 °C (95 °F). Each layer acts as a barrier against various epoxies and industrial chemicals. Can be worn as a liner under regular boots, or as a cover over shoes or boots. One size fits all.
Customers Who Bought This Also Bought