DuPont™ ProShield® 60 Lab Coats
Supplier: DuPont
Certificates
About this item
ProShield® 60 garments are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
- Laydown collar
- Open wrists
- Length extends below hips
- Front snap closure
- Two pockets
- Serged seams
Certifications: Thickness meets ASTM D751.
Ordering information: ProShield® 60 garments are made with a microporous film that is laminated to a spunbonded polypropylene substrate. Applications include automotive refinishing, waste cleanup and remediation. Lab coats offer front snap closures for easy donning and doffing. The front snap closure provides enhanced frontal protection. Additional features include laydown collar, open wrists, and two front pockets for holding writing instruments, small tools, or parts.
Specifications
- Catalog No:
- CA13452-318
- CA13452-320
- 10016-126
- CA13452-312
- CA13452-316
- CA13452-322
- CA13452-324
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory