SP Wilmad-LabGlass Reducing [ST] Adapters, SP Industries
Supplier: Avantor
Certificates
About this item
These adapters are manufactured from borosilicate glass.
- Top outer joint
- Bottom inner joint
Reducing and enlarging adapters allow connection of dissimilar Standard Taper joints. Outer joint at the top, inner joint at the bottom.
Customers Who Bought This Also Bought