SP Wilmad-LabGlass Reducing Adapters, Bushing Style, SP Industries
Supplier: Avantor
Certificates
About this item
This reducing adapter provides compact Standard Taper assemblies.
- Larger [ST] size on outside bottom
- Smaller [ST] size on top
The larger Standard Taper size is on the outside bottom of bushing while the smaller Standard Taper dimension is on top within the bore of bushing.
Customers Who Bought This Also Bought