Flame Resistant Deluxe Reflective Coverall with Max Comfort™ Material, Red
Catalog #: 10813-052
Supplier: MCR Safety
UL Listed
Specifications
- Length:Tall
- Color:Red
- Thickness:7 oz.
- Supplier No.:DC1RR38T
Specifications
About this item
Whether working in the rugged oil and gas industry or spending your day working near electrical hazards, stay safe and comfortable in MCR Safety's FR Gear made with Max Comfort™ fabrics.
- Silver reflective tape on legs, sleeves, and shoulders
- Two needle reinforced stitching on major seams for durability
- Heavy-duty two-way YKK® brass break-away zipper with Neomex® tape
- Multiple pockets on chest, back, hips, arm and leg
The DC1 coverall features a comfortable 88% cotton 12% Nylon blend that meets the NFPA 2112-2012 Standard for Flame Resistant Garments and NFPA 70E Standard for Electrical Safety in the Workplace. All MCR Safety's FR Garments are UL Certified and have a lifetime FR guarantee.
Size: 38T
Certifications: NFPA 2112, NFPA 70E, UL certified.
Customers Who Bought This Also Bought