Virtu-Clean™ Nitrile Examination Gloves
Supplier: High-Tech Conversions
New Product
Certificates
About this item
Virtu-Clean™ class 100 compatible powder-free nitrile examination gloves.
- Patient examination grade
- Non sterile
- Powder-free, with textured fingers
- Ambidextrous
- 100% nitrile; latex-free
- 12" length for added safety
Virtu-Clean™ ISO Class 5/Class 100 compatible gloves are ambidextrous, latex-free, and powder-free. Featuring a 12" cuff, these gloves are ideal for use in critical environments, as they will protect against the harmful effects of hazardous drugs.
Certifications: USP compliant. ISO Class 5/Class 100 compatible. Tested for use with Chemotherapy Drugs per ASTM D6978.
Packaging: Gloves are packaged 100 per bag, 10 bags per case.
Specifications
- Catalog No:
- 77507-154
- 77507-148
- 77507-146
- 77507-150
- 77507-152
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cleanroom class:
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- Finger thickness:
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
- 4.5 mil
Specifications
Customers Who Bought This Also Bought