About this item
Eppendorf rotors for centrifuge 5427 R redefine efficiency. Tailor your centrifugation with various capacities and speeds. Explore accessory options for enhanced versatility, ensuring precise sample processing for diverse applications.
- Precision-engineered for optimal sample separation
- All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121 °C, 20 minutes)
- Accessories (adaptors and buckets) further customize your centrifuge setup
- Aerosol-tight options available
Elevate your centrifugation experience with Eppendorf rotors crafted for centrifuge 5427 R. These precision rotors offer diverse capacities and speeds, providing flexibility for microcentrifugation or larger sample volumes. Accessories such as adaptors and buckets enhance customization for specific applications. Maximize efficiency with reliable sample separation, meeting the demands of molecular biology, genomics, and more. Explore the full range of options to tailor your centrifuge setup, achieving reproducible results and optimizing your laboratory workflow.
Specifications
- Catalog No:
- CA89239-150
- CA89239-148
- CA89239-142
- CA11028-848
- CA89239-134
- CA89239-138
- CA89239-136
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001
- Renewable Energy Certificate, ISO 14001