AssureTouch™ Fully Textured Latex Medical Examination Gloves, Powder-free
Supplier: Medicom
New Product
Certificates
About this item
Natural rubber latex offers maximum dexterity and tactility while providing reliable barrier protection. Fully textured for optimal grip.
- Fully textured
- Excellent manual dexterity
- Comfortable fit
- Optimal tactile sensitivity
These latex gloves conform closely to hands to provide maximum comfort, dexterity, and tactile sensitivity. They are fully textured for optimal grip and are puncture-resistant, as the latex material will stretch before it tears. This makes them an ideal choice when working with sharp instruments.
Certifications: ISO 11193-1 - type 1.
Packaging: Gloves are packaged 100 per box, 10 boxes per case (1000 total).
Specifications
- Catalog No:
- CA77321-834
- CA77321-838
- CA77321-836
- CA77321-842
- CA77321-840
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Latex
- Latex
- Latex
- Latex
- Latex
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile