Langenbeck Bone Saw
Catalog #: 470334-854
Supplier: AFTAB AHMED SE
Specifications
- Length:4.5"
- Description:Bone saw
- Material:Stainless steel
Specifications
About this item
Easily cut larger bones.
- Stainless steel
During large dissections you may encounter bones that are too big for bone cutters. This hand saw is an easy to use alternative to power saws.
The long blade and double row of teeth provide quick cutting.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Customers Who Bought This Also Bought