TruCLEAN® Sponge Blocks and Wipes, Perfex
Supplier: Perfex
Certificates
About this item
Polyfoam sponges and wipes are fabricated from a unique polyurethane foam that freely absorbs and holds liquids.
- Compatible with gamma, ETO, and autoclave sterilization up to 121 °C (250 °F) for 30 minutes
- Dampen sponge or wipe prior to autoclave sterilization
Hydro-Sorb wipes are specifically designed for use in controlled wet environments requiring rapid pickup of liquids. They are also ideal for general cleaning applications and quickly dry surfaces without leaving any residue. Hydro-Sorb wipes are made of unique hydrophilic, open cell polyurethane foam that is impervious to most chemicals and holds up to 15 times its weight in liquids.
They offer fast wet out, excellent wipe-dry, and fluid release. Chemical-resistant sponges and wipers are ideal for absorbing, retaining, and transferring liquids. Soft and pliable when dry. Non-linting and dimensionally stable.
Customers Who Bought This Also Bought