LD-100 System Easy-On Hoods, WORKLON®
Supplier: Superior Uniform
Certificates
About this item
Reusable, 100% polyester taffeta, cleanroom hoods feature an adjustable snap front closure, as well as a rear horizontal snap adjustment
Two sets of vertical snaps provide for a facemask attachment. Hood bib drapes across the shoulders. A color-coded hang loop denotes size. Hoods are gamma compatible and autoclavable.
Specifications
- Catalog No:
- 14227-836
- 14227-838
- 14227-834
- 14227-840
- 14227-842
- Garment application:
- Industrial, Cleanroom
- Industrial, Cleanroom
- Industrial, Cleanroom
- Industrial, Cleanroom
- Industrial, Cleanroom
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Fabric:
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester