VWR® Signature™ Frocks made with DuPont™ Tyvek® IsoClean® Material
Supplier: VWR International
Certificates
About this item
Frocks provide a high level of protection in cleanroom environments.
- High level of protection in cleanroom environments
- Chemically and biologically inert
- Low linting
- Static-free until laundered
These limited-use frocks are available in two styles. Frocks with covered zipper front feature tunneled elastic wrists and serged seams. These frocks are available with a bound neck. Frocks with five-snap front feature a bound neck and bound seams. All garments are pocketless.
Garments are chemically and biologically inert, low-linting, and static-free until laundered.
Specifications
- Catalog No:
- 80200-600
- 89127-316
- 89127-308
- 80200-604
- 80200-602
- 80200-608
- 80200-606
- 89128-788
- 80200-612
- 80200-610
- 89127-312
- 89125-608
- Garment application:
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
- Laboratory
Specifications
Related Information
Certificates
Search by Lot Number80200-606_P 80200-612_P 89125-608_P 89127-308_P 89127-316_P
BSE/TSE Certificates
Focus Safety Volume 1 Issue 1 Safety Essentials Volume 2
Product Literature
Customers Who Bought This Also Bought