ContactUsLinkComponent
OptiVISOR® Binocular Headband Magnifier
OptiVISOR® Binocular Headband Magnifier
Catalog #: 36968-007
Supplier:  Donegan Optical
OptiVISOR* Headband Magnifier
OptiVISOR® Binocular Headband Magnifier
Catalog #: 36968-007
Supplier:  Donegan Optical
Specifications
  • Magnification:
    2.75×
  • Cat. no.:
    36968-007
  • Supplier No.:
    DA-7 BLUE
Specifications
About this item
For close or precision work requiring visual accuracy
Continuous headband with dial adjustment for comfortable wear. Visor and lens pivot up and out of the way for normal vision when not in use. Fits over regular glasses. Magnifies with 15.2cm (6") focal length.
Customers Who Bought This Also Bought