Synthware Thermometer Adapter, Kemtech America
Catalog #: 60000-288
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- ST Joint:10/30
- Description:Synthware thermometer adapter
- Cat. no.:60000-288
- Supplier No.:A162440
Specifications
About this item
This adapter features a thermometer joint that is angled for use in a multiple neck flask.
- Angled joint
- Accommodates thermometer
The angled thermometer joint is size [ST] 10/30.
Customers Who Bought This Also Bought