Synthware Adapters with Mineral Oil Bubbler
Catalog #: 60000-696
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- ST Joint:24/40
- Cat. no.:60000-696
- Supplier No.:A402440
Specifications
About this item
This mineral oil bubbler has a 10 mm O.D.
- [ST] joint
- With hose connection
Features a standard taper joint and hose connection.
Customers Who Bought This Also Bought