ContactUsLinkComponent
DuPont™ ProShield® 60 Bib Aprons
DuPont™ ProShield® 60 Bib Aprons
Catalog #: CA13451-080
Supplier:  DuPont
ProShield* NexGen* Bib Aprons
DuPont™ ProShield® 60 Bib Aprons
Catalog #: CA13451-080
Supplier:  DuPont
Specifications
  • Garment application:
    Industrial,
    General Purpose
  • W×L:
    27×353/4"
  • Color:
    White
  • Cat. no.:
    CA13451-080
  • Supplier No.:
    NG273BWH0000
Specifications
About this item
ProShield® 60 garments are suitable for protection against non-hazardous particles, light liquid splash and aerosols in many industrial applications.
  • Neck loop
  • Waist ties
  • Bound seams
  • One size fit most
ProShield® 60 garments are made with a microporous film that is laminated to a spunbonded polypropylene substrate. Applications include automotive refinishing, waste cleanup and remediation.
These bib aprons provide enhanced frontal protection. Bound seams around neck and ties at waist allow for one size to fit most.
Certifications: Thickness meets ASTM D751.
Customers Who Bought This Also Bought