ContactUsLinkComponent
BD BBL™ Test Tube Agglutination Viewer, BD Diagnostics
  CA63020-066
 :  BD
undefined
BD BBL™ Test Tube Agglutination Viewer, BD Diagnostics
  CA63020-066
 :  BD
 :  420630
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Agglutination Viewer
  • Environmentally Preferable:
  • Cat. no.:
    CA63020-066
  • Supplier No.:
    420630
Ideal for use in blood typing, cross matching, flocculation, serodiagnostic tests, and other agglutinations
Viewer provides an easy and accurate reading of the undersurface of test tube contents. Sample contents are illuminated from above by a 5W bulb and from below by a magnifying mirror. Replacement bulbs available separately.