About this item
Save costs and deliver results in your hypoxia experiments. The CellXpert C170i CO₂ incubator with O₂ control helps you save money (no expandable parts like HEPA filters, UV lamps, etc.). Keep contaminants away with the 180 °C High-Temp Disinfection. Grow your cells consistently with fast recovery, uniform temperature and no vibration from a fan.
- Easy cleaning and disinfection: Seamless chamber, easy disassembling in 40 seconds, removable water tray
- Certified, effective contamination protection: 180 °C High temperature disinfection (HTD) with protocol, no internal fan-associated HEPA filters necessary
- Uniform temperature: Innovative heating concept with four temperature sensors at various locations verified at 27 locations inside the incubator (German DIN 12880:2007-05)
- Fast recovery after door opening: Temperature and CO₂ in less than five minutes without regulation overshoot
- Enhanced vibration and turbulence protection: No fan inside
- Save money: No internal expendable parts (e.g. fan-associated HEPA filters), significantly reduced gas consumption, up to 25% more usable space by fanless design and innovative insulation concept
- Stay flexible for the future: Upgrade your device in the laboratory later with various options
- Full control and easy documentation: Advanced user interface, two front USB-ports, customized tasks and alarms, VisioNize® user management and connected lab
Today's cell culture requires more than technical solutions to keep operational costs under control, optimize cell growth conditions, and efficiently prevent contamination.
The CellXpert® CO₂ incubator family was designed with the needs of modern cell culture labs in mind - to reduce time-to-market or publication with minimal efforts and resources. They are the perfect partner for demanding applications like assay development or culture of stem cells, primary cells, or 3D organoids and tissues.
O₂ Sensor: ZrO₂ (zirconium dioxide)
Humidity delivery: Water tray
Copper package: Chamber, water tray and four shelves
CO₂ uniformity: ±0.1%
Specifications
- Catalog No:
- CA77321-644
- CA77321-642
- CA76468-860
- CA77321-646
- CA77321-640
- CA77321-638
- CA76534-620
- CA76534-616
- CA76534-618
- CA76534-614
- CA77321-650
- CA77321-648
- Capacity:
- 170 L (6.0 ft³)
- 170 L (6.0 ft³)
- 170 L (6.0 ft³)
- 170 L (6.0 ft³)
- 170 L (6.0 ft³)
- 170 L (6.0 ft³)
- 170 L (6.0 ft³)
- 170 L (6.0 ft³)
- 170 L (6.0 ft³)
- 170 L (6.0 ft³)
- 170 L (6.0 ft³)
- 170 L (6.0 ft³)
- CO₂ range:
- 0.1 - 20%
- 0.1 - 20%
- 0.1 - 20%
- 0.1 - 20%
- 0.1 - 20%
- 0.1 - 20%
- 0.1 - 20%
- 0.1 - 20%
- 0.1 - 20%
- 0.1 - 20%
- 0.1 - 20%
- 0.1 - 20%
- Temperature uniformity:
- ±0.3 °C
- ±0.3 °C
- ±0.3 °C
- ±0.3 °C
- ±0.3 °C
- ±0.3 °C
- ±0.3 °C
- ±0.3 °C
- ±0.3 °C
- ±0.3 °C
- ±0.3 °C
- ±0.3 °C
- Temperature range:
- Ambient +4…50 °C
- Ambient +4…50 °C
- Ambient +4…50 °C
- Ambient +4…50 °C
- Ambient +4…50 °C
- Ambient +4…50 °C
- Ambient +4…50 °C
- Ambient +4…50 °C
- Ambient +4…50 °C
- Ambient +4…50 °C
- Ambient +4…50 °C
- Ambient +4…50 °C
- Int. dimensions:
- 53.9×44.5×69.2 cm (21.2×17.5×27.2")
- 53.9×44.5×69.2 cm (21.2×17.5×27.2")
- 53.9×44.5×69.2 cm (21.2×17.5×27.2")
- 53.9×44.5×69.2 cm (21.2×17.5×27.2")
- 53.9×44.5×69.2 cm (21.2×17.5×27.2")
- 53.9×44.5×69.2 cm (21.2×17.5×27.2")
- 53.9×44.5×69.2 cm (21.2×17.5×27.2")
- 53.9×44.5×69.2 cm (21.2×17.5×27.2")
- 53.9×44.5×69.2 cm (21.2×17.5×27.2")
- 53.9×44.5×69.2 cm (21.2×17.5×27.2")
- 53.9×44.5×69.2 cm (21.2×17.5×27.2")
- 53.9×44.5×69.2 cm (21.2×17.5×27.2")
- Exterior dimensions:
- 71.8×71.5×90.0 cm (28.3×28.1×35.4")
- 71.8×71.5×90.0 cm (28.3×28.1×35.4")
- 71.8×71.5×90.0 cm (28.3×28.1×35.4")
- 71.8×71.5×90.0 cm (28.3×28.1×35.4")
- 71.8×71.5×90.0 cm (28.3×28.1×35.4")
- 71.8×71.5×90.0 cm (28.3×28.1×35.4")
- 71.8×71.5×90.0 cm (28.3×28.1×35.4")
- 71.8×71.5×90.0 cm (28.3×28.1×35.4")
- 71.8×71.5×90.0 cm (28.3×28.1×35.4")
- 71.8×71.5×90.0 cm (28.3×28.1×35.4")
- 71.8×71.5×90.0 cm (28.3×28.1×35.4")
- 71.8×71.5×90.0 cm (28.3×28.1×35.4")
- Electrical:
- 100 - 127 V, 50/60 Hz (US/JP/South America)
- 100 - 127 V, 50/60 Hz (US/JP/South America)
- 100 - 127 V, 50/60 Hz (US/JP/South America)
- 100 - 127 V, 50/60 Hz (US/JP/South America)
- 100 - 127 V, 50/60 Hz (US/JP/South America)
- 100 - 127 V, 50/60 Hz (US/JP/South America)
- 100 - 127 V, 50/60 Hz (US/JP/South America)
- 100 - 127 V, 50/60 Hz (US/JP/South America)
- 100 - 127 V, 50/60 Hz (US/JP/South America)
- 100 - 127 V, 50/60 Hz (US/JP/South America)
- 100 - 127 V, 50/60 Hz (US/JP/South America)
- 100 - 127 V, 50/60 Hz (US/JP/South America)