Nonmetallic Laboratory Safety Cans, Justrite®
Catalog #: 56609-586
Supplier: Justrite
UL Listed
Specifications
- Capacity:3.8 L (1 gal.)
- Description:Oval Shelf Can, Stainless Steel Faucet
- Supplier No.:14169
Specifications
About this item
High-density polyethylene safety cans for use on shelves or in tilt stands have self-closing faucet for convenient, controlled dispensing of hazardous liquids. Fill spout has self-closing pressure relief cap and double mesh flame arrester. The 18.9L (5gal.) cans have built-in vacuum breakers. Oval-shaped safety can 56609-586 has stainless steel faucet and round can 56609-589 has brass faucet, both mounted 5cm (2") above bottom. Tilt can 56609-588 with top faucet rests on a sturdy metal cradle which measures 36.8x35.5cm (14½x14") at the base.
- Feature additional self-closing faucet that allows safe dispensing of hazardous liquids into test tubes, small flasks, and beakers
- Fill spout has self-closing pressure relief cap and double mesh flame arrester
- Current-carrying carbon insert imbedded in the can provides proper grounding
- 5cm (2") with faucet mounted above the bottom allows dispensing without lifting
- 18.9L (5gal.) cans have built-in vacuum breaker
- FM approved
Customers Who Bought This Also Bought