PROSAT® Presaturated Wipers in SatBox™ Containers, Contec®
Supplier: Contec
Certificates
About this item
Premoistened knitted polyester wipers and nonwoven polypropylene wipers are saturated with various solvent blends.
- Made from high-density polypropylene
- Compatible with various blends of isopropyl alcohol, ethanol, and acetone
- Reduces solvent evaporation
Polynit Heatseal wipers are standard weight, textured 100% polyester interlock knit with nonabrasive, heatsealed edges. They are abrasion- and chemical-resistant. Economical nonwoven wipers are made of 100% meltblown polypropylene. PROSAT® wipers are ideal for wiping process tools and other equipment, as well as many other cleaning applications in critical environments. Color: white.
Ordering information: Wipers are packaged in foil pouches except for wipers 33501-490, which are packaged in resealable pouches for easy dispensing and to preserve cleanliness and solvent saturation levels. Each case includes one SatBox™ container, except for refill wipers 33501-828. Pouches are individually lot-numbered for traceability.
Related Information
Certificates
Search by Lot Number33501-490_812127-0-0-1 33501-828_699795-0-0-1 33501-828_716143-0-0-1 33501-828_718281-0-0-1 33501-828_741232-0-0-1
Conformity declaration
More
VWR GHS Compliance Letter (US)
Additional Product Details
Customers Who Bought This Also Bought